Få mere at vide
L-Leucine-2-d1, CDN
Artikelnummer. 30311137
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311137 0.1 g 0.1g
30318535 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311137 Leverandør CDN Leverandørnr. CDND59370.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Leucine-2-D
CAS 89836-93-1
Molekylær formel C6 D H12 N O2
Mængde 0.1 g
Synonym L-Leucine-2-d (9 CI, ACI), L-[2-2 H]Leucine, L-Leucine-2-D, (S)-4-Methyl-2-aminopentanoic-2-d acid, (2 S)-4-Methyl-2-aminopentanoic-2-d acid, L-Leucine-α-d
InChI formel InChI=1 S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i5D
SMIL [2 H][C@](N)(CC(C)C)C(=O)O
IUPAC navn (2 S)-2-amino-2-deuterio-4-methylpentanoic acid
Molekylvægt (g/mol) 132.1791
Formel vægt 132.1009 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity