Få mere at vide
L-Lactic Acid-d3 Sodium Salt, TRC
Artikelnummer. 30479919
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479919 2.5 mg
30478893 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479919 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1135032.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 285979-84-2
Molekylær formel C3 2H3 H2 O3 . Na
Molekylvægt (g/mol) 115.08
Synonym Propanoic-3,3,3-d3 acid, 2-hydroxy-, monosodium salt, (2S)- (9CI), L-(+)-Lactic acid-d3 sodium salt, Sodium S-lactate-d3, Sodium (2S)-2-hydroxypropanoate-3,3,3-d3, (S)-2-Hydroxypropionic-3,3,3-d3 acid sodium salt, Sodium (S)-lactate-3,3,3-d3, Sodium L-(+)-lactate-d3, Sodium L-lactate-3,3,3-D3
IUPAC navn sodium;(2S)-3,3,3-trideuterio-2-hydroxypropanoate
SMIL [Na+].[2H]C([2H])([2H])[C@H](O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale L-Lactic Acid-d3 Sodium Salt
Mængde 2.5 mg
InChI formel InChI=1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i1D3;
Formel vægt 115.0325