Learn More
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
Catalog Number p-3757422
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
79-33-4, 7732-18-5
C3H6O3
90.078
MFCD00064266
JVTAAEKCZFNVCJ-REOHCLBHSA-N
l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
107689
CHEBI:422
(2S)-2-hydroxypropanoic acid
CC(C(=O)O)O
79-33-4, 7732-18-5
C3H6O3
90.078
MFCD00064266
JVTAAEKCZFNVCJ-REOHCLBHSA-N
l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
107689
CHEBI:422
(2S)-2-hydroxypropanoic acid
CC(C(=O)O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless
Beilstein 03, IV, 633
Fieser 15,181
Merck Index 15, 5385
Formula Weight 90.08
Percent Purity ≥18%
Density 1.0500g/mL
Quantity 1 L
Packaging Glass bottle
Physical Form Liquid
Chemical Name or Material L(+)-Lactic acid
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.