Få mere at vide
L-Kynurenine, TRC
Artikelnummer. 30407604
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30407604 100 mg
30404733 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30407604 Leverandør Toronto Research Chemicals Leverandørnr. TRCK661005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Kynurenine
CAS 2922-83-0
Renhedsgrad noter HPLC
Molekylær formel C10 H12 N2 O3
Mængde 100 mg
Synonym Benzenebutanoic acid, α,2-diamino-γ-oxo-, (αS)-, Alanine, 3-anthraniloyl-, L- (8CI), Benzenebutanoic acid, α,2-diamino-γ-oxo-, (S)-, (αS)-α,2-Diamino-γ-oxobenzenebutanoic acid, (2S)-4-(2-Aminophenyl)-2-azaniumyl-4-oxobutanoate, 3-Anthraniloylalanine, Kynurenin, Kynurenine, L-3-(o-Aminobenzoyl)alanine, L-Kynurenine, (S)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid
InChI formel InChI=1S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8H,5,11-12H2,(H,14,15)/t8-/m0/s1
SMIL N[C@@H](CC(=O)c1ccccc1N)C(=O)O
IUPAC navn (2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid
Molekylvægt (g/mol) 208.21
Formel vægt 208.0848
Procent renhed >95