Få mere at vide
L-Isoleucine-2-d1, TRC
Artikelnummer. 30179350
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179350 10 mg
30179230 50 mg
30179323 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179350 Leverandør Toronto Research Chemicals Leverandørnr. TRCI82017610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 127290-60-2
Molekylær formel C6 D H12 N O2
Molekylvægt (g/mol) 132.179
Synonym L-Isoleucine-2-d1, L-Isoleucine-2-d (9CI, ACI), L-Isoleucine-2-D, (2S,3S)-2-Amino-3-methylpentanoic-2-d acid
IUPAC navn (2S,3S)-2-amino-2-deuterio-3-methylpentanoic acid
SMIL [2H][C@](N)([C@@H](C)CC)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Isoleucine-2-d1
Mængde 10 mg
InChI formel InChI=1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1/i5D
Formel vægt 132.101