Få mere at vide
L-Iduronic Acid Sodium Salt, TRC
Artikelnummer. 17957264
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17957264 50 mg
17927644 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17957264 Leverandør Toronto Research Chemicals Leverandørnr. TRCI25200050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61199-83-5
Molekylær formel C6H9NaO7
Molekylvægt (g/mol) 216.12
Synonym L-Iduronic acid, monosodium salt (9CI), Sodium L-iduronate, L-Iduronic acid sodium salt
SMIL [Na+].O[C@@H](C=O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale L-Iduronic Acid, Sodium Salt
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C6H10O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h1-5,8-11H,(H,12,13);/q;+1/p-1/t2-,3+,4-,5+;/m0./s1
Procent renhed >95%