missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
L-Homoserine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-Homoserine |
| CAS | 672-15-1 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C4 H9 N O3 |
| Mængde | 1 g |
| Synonym | 2(3H)-Furanone, 3-aminodihydro-, hydrobromide (1:1) (ACI), 2(3H)-Furanone, 3-aminodihydro-, hydrobromide (9CI), 2(3H)-Furanone, 3-aminodihydro-, hydrobromide, (±)- (ZCI), (±)-3-Aminodihydro-2(3H)-furanone hydrobromide, 2-Amino-4-butyrolactone hydrobromide, 2-Aminobutyrolactone hydrobromide, 3-Aminodihydro-2(3H)-furanone hydrobromide, 3-Aminooxolan-2-one hydrobromide, DL-Homoserine lactone hydrobromide, Alpha-amino-gamma-butyrolactone hydrobromide, Homoserine lactone hydrobromide, NSC 41120, α-Amino-γ-butyrolactone hydrobromide |
| InChI formel | InChI=1S/C4H9NO3/c5-3(1-2-6)4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m0/s1 |
| SMIL | N[C@@H](CCO)C(=O)O |
| IUPAC navn | (2S)-2-amino-4-hydroxybutanoic acid |
| Molekylvægt (g/mol) | 119.12 |
| Vis mere |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?