Få mere at vide
L-Homocysteine, TRC
Artikelnummer. 30136641
Skift visning
Mængde:
25 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136641 50 mg
30136886 500 mg
30004725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136641 Leverandør Toronto Research Chemicals Leverandørnr. TRCH59195050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6027-13-0
Molekylær formel C4 H9 N O2 S
Molekylvægt (g/mol) 135.1848
Synonym L-Homocysteine, Butyric acid, 2-amino-4-mercapto-, L- (8CI), (S)-2-Amino-4-mercaptobutanoic acid, (S)-Homocysteine, 2-Amino-4-mercapto-L-butyric acid, 2-Amino-4-mercaptobutyric acid, Butanoic acid, 2-amino-4-mercapto-, (S)-, Homocysteine, NSC 43117
IUPAC navn (2S)-2-amino-4-sulfanylbutanoic acid
SMIL N[C@@H](CCS)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Homocysteine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C4H9NO2S/c5-3(1-2-8)4(6)7/h3,8H,1-2,5H2,(H,6,7)/t3-/m0/s1
Mængde 50 mg
Formel vægt 135.0354
Procent renhed >95%