Få mere at vide
L-Homocysteine-d4, TRC
Artikelnummer. 30136250
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136250 25 mg -
17927829 2.5 mg 2.5mg
17988949 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30136250 Leverandør Toronto Research Chemicals Leverandørnr. TRCH59195225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 331665-12-4
Molekylær formel C4H5D4NO2S
Molekylvægt (g/mol) 139.21
Synonym Homocysteine-d4, NSC 43117-d4, (S)-2-Amino-4-mercaptobutanoic Acid-d4, (S)-2-Amino-4-mercaptobutanoic Acid-d4, (S)-Homocysteine-d4, 2-Amino-4-mercapto-L-butyric Acid-d4, 2-Amino-4-mercaptobutyric Acid-d4, L-2-Amino-4-mercaptobutyric Acid-d4;
IUPAC navn (2S)-2-amino-3,3,4,4-tetradeuterio-4-sulfanylbutanoic acid
SMIL [2H]C([2H])(S)C([2H])([2H])[C@H](N)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Homocysteine-d4
InChI formel InChI=1S/C4H9NO2S/c5-3(1-2-8)4(6)7/h3,8H,1-2,5H2,(H,6,7)/t3-/m0/s1/i1D2,2D2
Mængde 25 mg
Formel vægt 139.0605