missing translation for 'onlineSavingsMsg'
Learn More

L-Homoarginine Hydrochloride, TRC

Product Code. 30137182
Change view
Click to view available options
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30137182 5 g
30135746 10 g
30004406 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137182 Supplier Toronto Research Chemicals Supplier No. TRCH5850005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1483-01-8
Molekylær formel C7 H16 N4 O2 . Cl H
Molekylvægt (g/mol) 224.69
Synonym L-Lysine, N6-(aminoiminomethyl)-, hydrochloride (1:1) (ACI), L-Lysine, N6-(aminoiminomethyl)-, monohydrochloride (9CI), Lysine, N6-amidino-, monohydrochloride, L- (8CI), (S)-2-Amino-6-guanidinohexanoic acid hydrochloride, L-Homoarginine hydrochloride, NSC 145416, (2S)-2-Amino-6-guanidinohexanoic acid hydrochloride, N6-Amidino-L-lysine monohydrochloride, N6-(Aminoiminomethyl)-L-lysine monohydrochloride
IUPAC navn (2S)-2-amino-6-carbamimidamidohexanoic acid;hydrochloride
SMIL Cl.N[C@@H](CCCCNC(=N)N)C(=O)O

Specifications

Kemisk navn eller materiale L-Homoarginine, Hydrochloride
Renhedsgrad noter HPLC
InChI formel InChI=1S/C7H16N4O2.ClH/c8-5(6(12)13)3-1-2-4-11-7(9)10;/h5H,1-4,8H2,(H,12,13)(H4,9,10,11);1H/t5-;/m0./s1
Mængde 5 g
Formel vægt 224.104
Procent renhed >95%
Forsendelsestilstand Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.