Få mere at vide
L-Glutamine-2,3,3,4,4-d5, TRC
Artikelnummer. 30406952
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30406952 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406952 Leverandør Toronto Research Chemicals Leverandørnr. TRCG59700510MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Glutamine-2,3,3,4,4-d5
CAS 14341-78-7
Molekylær formel C5 D5 H5 N2 O3
Mængde 10 mg
Synonym (S)-2,5-Diamino-5-oxopentanoic Acid-d5, 2-Aminoglutaramic Acid-d5, Acustasin-d5, Aesgen 14-d5, Cebrogen-d5, Glumin-d5, L-2-Aminoglutaramidic Acid-d5, L-Glutamic Acid γ-αmide-d5, Levoglutamide-d5, NSC 27421-d5, Saforis-d5, Stimulina-d5, γ-Glutamine-d5
InChI formel InChI=1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1/i1D2,2D2,3D
SMIL [2H]C([2H])(C(=O)N)C([2H])([2H])[C@]([2H])(N)C(=O)O
IUPAC navn (2S)-2,5-diamino-2,3,3,4,4-pentadeuterio-5-oxopentanoic acid
Molekylvægt (g/mol) 151.18
Formel vægt 151.1005