Få mere at vide
L-glutamin dehydrogenase, MP Biomedicals™
Artikelnummer. 11463701 Shop alle MP Biomedicals produkter
Skift visning
Mængde:
100 mg
1 g
Pakningsstørrelse:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11463701 100 mg 100mg
11483701 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11463701 Leverandør MP Biomedicals Leverandørnr. 0215118980
Tekniske data
Molekylær formel C6H12O6
CAS 9029-12-3
Mængde 100 mg
Molekylvægt (g/mol) 180.16
Synonym cerebrose
Form Overskyet affjedring