Få mere at vide
L-Glutamic Acid Monoammonium Salt, TRC
Artikelnummer. 30240745
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240745 10 g
30240955 5 g
30240702 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240745 Leverandør Toronto Research Chemicals Leverandørnr. TRCG59714010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7558-63-6
Molekylær formel C5H9NO4 . NH3
Molekylvægt (g/mol) 164.16
Synonym Ammonium Glutamate, E 624, Glutamic Acid Monoammonium Salt, Monoammonium L-Glutamate, Monoammonium Glutamate, L-Glutamic Acid Ammonium Salt;
IUPAC navn azanium;(2S)-2-amino-5-hydroxy-5-oxopentanoate
SMIL [NH4+].N[C@@H](CCC(=O)O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale L-Glutamic Acid Monoammonium Salt
InChI formel InChI=1S/C5H9NO4.H3N/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);1H3/t3-;/m0./s1
Mængde 10 g
Formel vægt 164.0797