Få mere at vide
L-Glutamic Acid gamma-p-Nitroanilide, TRC
Artikelnummer. 30405252
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405252 5 g
30403206 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30405252 Leverandør Toronto Research Chemicals Leverandørnr. TRCG5970505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Glutamic Acid gamma-p-Nitroanilide
CAS 7300-59-6
Molekylær formel C11 H13 N3 O5
Mængde 5 g
Synonym N-(4-Nitrophenyl)-L-glutamine, 2-Amino-4'-nitroglutaranilic Acid, L-Glutamic Acid γ-p-Nitroanilide, L-Glutamyl-4-nitroanilide, L-γ-Glutamyl-p-nitroanilide, γ-Glutamyl-p-nitroanilide, γ-L-Glu-p-nitroanilide, γ-L-Glutamyl-p-nitroanilide
InChI formel InChI=1S/C11H13N3O5/c12-9(11(16)17)5-6-10(15)13-7-1-3-8(4-2-7)14(18)19/h1-4,9H,5-6,12H2,(H,13,15)(H,16,17)/t9-/m0/s1
SMIL N[C@@H](CCC(=O)Nc1ccc(cc1)[N+](=O)[O-])C(=O)O
IUPAC navn (2S)-2-amino-5-(4-nitroanilino)-5-oxopentanoic acid
Molekylvægt (g/mol) 267.24
Formel vægt 267.0855