missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid diethyl ester hydrochloride, 98%, ACROS Organics™

Brand:  Acros Organics 459260250

Additional Details : CAS Number : 1118-89-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15482262

  • 601.11 DKK / 25g


Estimated Shipment
12-10-2020
Add to basket

Specifications

Chemical Identifiers

1118-89-4
239.7
l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
diethyl (2S)-2-aminopentanedioate;hydrochloride
C9H17NO4·HCl
WSEQLMQNPBNMSL-FJXQXJEOSA-N
73960
CCOC(=O)CCC(C(=O)OCC)N.Cl

Specifications

L-Glutamic acid diethyl ester hydrochloride
108.0°C to 110.0°C
1118-89-4
l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
WSEQLMQNPBNMSL-FJXQXJEOSA-N
diethyl (2S)-2-aminopentanedioate;hydrochloride
73960
98%
Authentic
25g
C9H17NO4·HCl
(5% in water) Clear colorless
CCOC(=O)CCC(C(=O)OCC)N.Cl
239.7
239.7
Videos
Documents