missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid diethyl ester hydrochloride, 98%, Thermo Scientific™

Catalog Number p-7077784
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1118-89-4
C9H17NO4·HCl
239.7
WSEQLMQNPBNMSL-FJXQXJEOSA-N
l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
73960
diethyl (2S)-2-aminopentanedioate;hydrochloride
CCOC(=O)CCC(C(=O)OCC)N.Cl
This item is not returnable. View return policy
1118-89-4
C9H17NO4·HCl
239.7
WSEQLMQNPBNMSL-FJXQXJEOSA-N
l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
73960
diethyl (2S)-2-aminopentanedioate;hydrochloride
CCOC(=O)CCC(C(=O)OCC)N.Cl
Melting Point 108.0°C to 110.0°C
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information (5% in water) Clear colorless
Formula Weight 239.7
Percent Purity 98%
Chemical Name or Material L-Glutamic acid diethyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'