Få mere at vide
L-Glutamic Acid alpha-tert-Butyl Ester, TRC
Artikelnummer. 30004990
Skift visning
Mængde:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004990 25 g
30006336 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004990 Leverandør Toronto Research Chemicals Leverandørnr. TRCG59698025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 45120-30-7
Molekylær formel C9 H17 N O4
Molekylvægt (g/mol) 203.24
Synonym L-Glutamic Acid 1-(1,1-Dimethylethyl) Ester, L-Glutamic Acid tert-Butyl Ester, α-tert-Butyl L-Glutamate;
IUPAC navn (4S)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMIL CC(C)(C)OC(=O)[C@@H](N)CCC(=O)O
Tekniske data
Kemisk navn eller materiale L-Glutamic Acid alpha-tert-Butyl Ester
Renhedsgrad noter HPLC
InChI formel InChI=1S/C9H17NO4/c1-9(2,3)14-8(13)6(10)4-5-7(11)12/h6H,4-5,10H2,1-3H3,(H,11,12)/t6-/m0/s1
Mængde 25 g
Formel vægt 203.1158
Procent renhed >95%