missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
L-Glutamic acid Alpha-amide, TRC
Specifications
Specifications
| Kemisk navn eller materiale | L-Glutamic acid alpha-amide |
| CAS | 636-65-7 |
| Molekylær formel | C5H10N2O3 |
| Mængde | 100 mg |
| Synonym | L-isoglutamine, 4-amino-L-glutaramic acid, (4S)-4,5-Diamino-4-oxo-pentanoic acid |
| InChI formel | InChI=1S/C5H10N2O3/c6-3(5(7)10)1-2-4(8)9/h3H,1-2,6H2,(H2,7,10)(H,8,9)/t3-/m0/s1 |
| SMIL | N[C@@H](CCC(=O)O)C(=O)N |
| IUPAC navn | (4S)-4,5-diamino-5-oxopentanoic acid |
| Molekylvægt (g/mol) | 146.14 |
| Formel vægt | 146.0691 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?