missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Plastic Bottle
Specifications
Melting Point | 189°C to 192°C (decomposition) |
Linear Formula | HOOCCH2CH2CH(NH2)COOH |
Beilstein | 1723801 |
Packaging | Plastic Bottle |
Formula Weight | 147.13 |
Percent Purity | 98.5 to 101.5% |
Density | 1.525 |
Quantity | 5 kg |
Specific Gravity | 1.525 |
Physical Form | Crystalline Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'