missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
L-Glutamic acid 5-tert-butyl ester is used as a fine chemical intermediate, pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Glutamic acid 5-tert-butyl ester is used as a fine chemical intermediate, pharmaceutical intermediate.
Solubility
Sparingly soluble in water; practically insoluble in ethanol or ether.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
L-Glutamic acid 5-tert-butyl ester is used as a fine chemical intermediate, pharmaceutical intermediate.
Solubility
Sparingly soluble in water; practically insoluble in ethanol or ether.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Beilstein | 2049031 |
Synonym | h-glu otbu-oh, l-glutamic acid 5-tert-butyl ester, h-glu o-t-bu-oh, 2s-2-amino-5-tert-butoxy-5-oxopentanoic acid, l-gamma-t-butyl glutamate, l-glutamic acid,5-1,1-dimethylethyl ester, 5-tert-butyl l-glutamate, l-glutamic acid, 5-1,1-dimethylethyl ester, glu otbu, glu tbu |
Solubility Information | Sparingly soluble in water; practically insoluble in ethanol or ether. |
Formula Weight | 203.24 |
Percent Purity | 98% |
Quantity | 1 g |
Physical Form | Solid |
Chemical Name or Material | L-Glutamic acid 5-tert-butyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'