missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid 5-ethyl ester, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
L-Glutamic acid 5-ethyl ester is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water 100 mg/ml.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
L-Glutamic acid 5-ethyl ester is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water 100 mg/ml.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | 179°C (decomposition) |
Color | White |
Solubility Information | Soluble in water 100mg/ml. |
IUPAC Name | 2-amino-5-ethoxy-5-oxopentanoic acid |
Formula Weight | 175.18 |
Percent Purity | 95% |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | L-Glutamic acid 5-ethyl ester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'