missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid-13C5, TRC

Product Code. 30010070
Change view
Click to view available options
Mængde:
2.5 mg
5 mg
10 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30010070 2.5 mg 2.5mg
30004382 5 mg -
30008403 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010070 Supplier Toronto Research Chemicals Supplier No. TRCG5969612.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55443-55-5
Molekylær formel 13C5H9NO4
Molekylvægt (g/mol) 152.09
Synonym L-Glutamic-1,2,3,4,5-13C5 Acid
SMIL N[13C@@H]([13CH2][13CH2][13C](O)=O)[13C](O)=O

Specifications

Kemisk navn eller materiale L-Glutamic Acid-13C5
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1/i1+1,2+1,3+1,4+1,5+1
Formel vægt 152.0699
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.