missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 1-methyl ester, 98%, Thermo Scientific Chemicals

Catalog Number p-7054855
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6384-08-3
MFCD00171677
This item is not returnable. View return policy
6384-08-3
MFCD00171677

L-Glutamic acid 1-methyl ester nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 145°C (decomposition)
Color White
Molecular Formula C8H12N2
Beilstein 1707273
Synonym 2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4
Solubility Information Soluble in water.
InChI Key BWAPJIHJXDYDPW-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1N)C)N
IUPAC Name 2,5-dimethylbenzene-1,4-diamine
Molecular Weight (g/mol) 136.20
PubChem CID 160824
ChEBI CHEBI:53613
Formula Weight 161.16
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material L-Glutamic acid 1-methyl ester
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'