Få mere at vide
L-Glutamic-2,4,4-d3 Acid, CDN
Artikelnummer. 30327697
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327697 0.1 g 0.1g
30318822 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327697 Leverandør CDN Leverandørnr. CDND80100.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Glutamic-2,4,4-d3 Acid
CAS 203805-84-9
Molekylær formel C5 D3 H6 N O4
Mængde 0.1 g
Synonym L-Glutamic-2,4,4-d3 acid (9 CI, ACI), L-Glutamic Acid-2,4,4-D3, (2 S)-2-Aminopentanedioic-2,4,4-d3 acid, L-Glutamic Acid-d3
InChI formel InChI=1 S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1/i2D2,3 D
SMIL [2 H]C([2 H])(C[C@]([2 H])(N)C(=O)O)C(=O)O
IUPAC navn (2 S)-2-amino-2,4,4-trideuteriopentanedioic acid
Molekylvægt (g/mol) 150.1478
Formel vægt 150.072 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity