missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Galactose, 98%

Catalog Number p-7025780
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
15572-79-9
C6H12O6
180.156
MFCD00063833
GZCGUPFRVQAUEE-DPYQTVNSSA-N
aldehydo-l-galactose, 2s,3r,4r,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-galacto-hexose, unii-s93uii1dw8, s93uii1dw8, galactose, l
84996
CHEBI:37617
(2S,3R,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O
This item is not returnable. View return policy
15572-79-9
C6H12O6
180.156
MFCD00063833
GZCGUPFRVQAUEE-DPYQTVNSSA-N
aldehydo-l-galactose, 2s,3r,4r,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-galacto-hexose, unii-s93uii1dw8, s93uii1dw8, galactose, l
84996
CHEBI:37617
(2S,3R,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 163°C to 165°C
Quantity 100 mg
Beilstein 1724622
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material L-(-)-Galactose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'