Få mere at vide
L-(+)-Ergothioneine, TRC
Artikelnummer. 30004412
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004412 25 mg
17959644 5 mg
17987624 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004412 Leverandør Toronto Research Chemicals Leverandørnr. TRCE60000025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 497-30-3
Molekylær formel C9 H15 N3 O2 S
Molekylvægt (g/mol) 229.3
Synonym 1H-Imidazole-4-ethanaminium, α-carboxy-2,3-dihydro-N,N,N-trimethyl-2-thioxo-, inner salt, (αS)-, 1H-Imidazole-4-ethanaminium, α-carboxy-2,3-dihydro-N,N,N-trimethyl-2-thioxo-, hydroxide, inner salt, (S)-, Ammonium, [1-carboxy-2-(2-mercaptoimidazol-4-yl)ethyl]trimethyl-, hydroxide, inner salt, L-(+)- (8CI), (2S)-3-(2-Sulfanyl-1H-imidazol-4-yl)-2-(trimethylazaniumyl)propanoate, (S)-5-(2-Carboxy-2-(trimethylammonio)ethyl)-1H-imidazole-2-thiolate, 1H-Imidazole-4-ethanaminium, α-carboxy-2,3-dihydro-N,N,N-trimethyl-2-thioxo-, inner salt, (S)-, 2-Mercapto-N,N-dimethylhistidine methyl ester betaine, 2-Mercaptohistidine betaine, 2-Mercaptohistidine trimethylbetaine, Ergothioneine, Ergothionine, Histidine, 2-mercapto-, trimethylbetaine, Histidine, 2-mercapto-N,N-dimethyl-, betaine, L-Ergothioneine, NSC 7175, Phytothioneine, Sympectothion, Thiasine, Thioneine, Thiotane, thiotaine, L-(+)-Ergothioneine
IUPAC navn (2S)-3-(2-sulfanylidene-1,3-dihydroimidazol-4-yl)-2-(trimethylazaniumyl)propanoate
SMIL C[N+](C)(C)[C@@H](CC1=CNC(=S)N1)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale L-(+)-Ergothioneine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C9H15N3O2S/c1-12(2,3)7(8(13)14)4-6-5-10-9(15)11-6/h5,7H,4H2,1-3H3,(H2-,10,11,13,14,15)/t7-/m0/s1
Mængde 25 mg
Formel vægt 229.0885
Procent renhed >95%