Få mere at vide
L-(+)-Ergothioneine-d3, TRC
Artikelnummer. 30376907
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30376907 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376907 Leverandør Toronto Research Chemicals Leverandørnr. TRCE60000210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Ergothioneine N-methyl-d3
CAS 1356933-89-5
Molekylær formel C9 2H3 H12 N3 O2 S
Mængde 10 mg
Synonym 1H-Imidazole-4-ethanaminium, α-carboxy-2,3-dihydro-N,N-dimethyl-N-(methyl-d3)-2-thioxo-, inner salt, (αS)-, (2S)-2-[Dimethyl(trideuteriomethyl)azaniumyl]-3-(2-sulfanylidene-1,3-dihydroimidazol-4-yl)propanoate, (αS)-α-Carboxy-2,3-dihydro-N,N-dimethyl-N-(methyl-d3)-2-thioxo-1H-imidazole-4-ethanaminium inner salt, L-(+)-Ergothioneine-d3, L-Ergothioneine-d3, L-Ergothioneine-D3 (N-methyl-D3), (S)-[1-Carboxy-2-(2-mercaptoimidazol-4-yl)ethyl]dimethyl(methyl-d3)ammonium
InChI formel InChI=1S/C9H15N3O2S/c1-12(2,3)7(8(13)14)4-6-5-10-9(15)11-6/h5,7H,4H2,1-3H3,(H2-,10,11,13,14,15)/t7-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])[N+](C)(C)[C@@H](CC1=CNC(=S)N1)C(=O)[O-]
IUPAC navn (2S)-2-[dimethyl(trideuteriomethyl)azaniumyl]-3-(2-sulfanylidene-1,3-dihydroimidazol-4-yl)propanoate
Molekylvægt (g/mol) 232.32
Formel vægt 232.1073