Få mere at vide
L-DOPA-2,5,6-d3, TRC
Artikelnummer. 30379195
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30379195 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379195 Leverandør Toronto Research Chemicals Leverandørnr. TRCD533752250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-DOPA-2,5,6-d3
CAS 53587-29-4
Renhedsgrad noter HPLC
Molekylær formel C9 2H3 H8 N O4
Mængde 250 mg
Synonym L-Tyrosine-2,3,6-d3, 5-hydroxy- (9CI, ACI), 5-Hydroxy-L-tyrosine-2,3,6-d3 (ACI), L-Dihydroxyphenylalanine-2H3, (2S)-2-Amino-3-(3,4-dihydroxyphenyl-2,5,6-d3)propanoic acid, Levodopa-d3, Levodopa-D3 (phenyl-D3), L-Dopa-2,5,6-d3
InChI formel InChI=1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m0/s1/i1D,2D,4D
SMIL [2H]c1c([2H])c(C[C@H](N)C(=O)O)c([2H])c(O)c1O
IUPAC navn (2S)-2-amino-3-(2,3,6-trideuterio-4,5-dihydroxyphenyl)propanoic acid
Molekylvægt (g/mol) 200.21
Formel vægt 200.0876
Procent renhed >95