Få mere at vide
L-Dopa-2,5,6-d3, CDN
Artikelnummer. 30318045
Skift visning
Mængde:
0.5 g
Pakningsstørrelse:
0.5g
Artikelnummer. Mængde unitSize
30318045 0.5 g 0.5g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318045 Leverandør CDN Leverandørnr. CDND15700.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-DOPA-2,5,6-d3
CAS 53587-29-4
Molekylær formel C9 2H3 H8 N O4
Mængde 0.5 g
InChI formel InChI=1 S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12 H,3,10H2,(H,13,14)/t6-/m0/s1/i1D,2 D,4 D
SMIL [2 H]c1c([2 H])c(C[C@H](N)C(=O)O)c([2 H])c(O)c1O
IUPAC navn (2 S)-2-amino-3-(2,3,6-trideuterio-4,5-dihydroxyphenyl)propanoic acid
Molekylvægt (g/mol) 200.21
Formel vægt 200.0876 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity