Få mere at vide
L-Dopa-2,5,6,alpha,beta,beta-d6, CDN
Artikelnummer. 30327367
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327367 0.01 g
30313281 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327367 Leverandør CDN Leverandørnr. CDND78480.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Levodopa-D6
CAS 713140-75-1
Molekylær formel C9 D6 H5 N O4
Mængde 0.01 g
Synonym L-Tyrosine-α,β,β,2,3,6-d6, 5-hydroxy- (9 CI), 5-Hydroxy-L-tyrosine-α,β,β,2,3,6-d6, L-Dihydroxyphenylalanine-2H6, (2 S)-2-Amino-3-(3,4-dihydroxyphenyl-2,5,6-d3)propanoic-2,3,3-d3 acid, Levodopa-d6, Levodopa-D6, L-Dopa-d6, L-Dopa-2,5,6,α,β,β-d6
InChI formel InChI=1 S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12 H,3,10H2,(H,13,14)/t6-/m0/s1/i1D,2 D,3D2,4 D,6 D
SMIL [2 H]c1c([2 H])c(c([2 H])c(O)c1O)C([2 H])([2 H])[C@]([2 H])(N)C(=O)O
IUPAC navn (2 S)-2-amino-2,3,3-trideuterio-3-(2,3,6-trideuterio-4,5-dihydroxyphenyl)propanoic acid
Molekylvægt (g/mol) 203.2249
Formel vægt 203.1065 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity