missing translation for 'onlineSavingsMsg'
Learn More

L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7040614
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
5988-19-2
C5H6N2O4
158.113
MFCD00085339
UFIVEPVSAGBUSI-REOHCLBHSA-N
l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
439216
CHEBI:17025
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
C1C(NC(=O)NC1=O)C(=O)O
This item is not returnable. View return policy
5988-19-2
C5H6N2O4
158.113
MFCD00085339
UFIVEPVSAGBUSI-REOHCLBHSA-N
l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
439216
CHEBI:17025
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
C1C(NC(=O)NC1=O)C(=O)O

L-Dihydroorotic acid a metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Dihydroorotic acid a metabolite.

Solubility
Soluble in water (partly), and dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 254°C to 255°C (decomposition)
Quantity 1 g
Solubility Information Soluble in water (partly),and dimethyl formamide.
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material L-Dihydroorotic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'