Få mere at vide
L-Deoxyalliin, TRC
Artikelnummer. 30006971
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006971 10 mg
30005745 100 mg
30003964 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006971 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23218010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21593-77-1
Molekylær formel C6 H11 N O2 S
Molekylvægt (g/mol) 161.22
Synonym S-2-propen-1-yl-L-Cysteine, Alanine, 3-(allylthio)- (6CI,7CI), Alanine, 3-(allylthio)-, L- (8CI), L-Cysteine, S-2-propenyl- (9CI), (+)-S-Allylcysteine, 3-(Allylthio)-L-alanine, Deoxyalliin, NSC 96449, S-(2-Propenyl)-L-cysteine, S-2-Propenylcysteine, S-Allyl-L-cysteine, S-Allylcysteine
IUPAC navn (2R)-2-amino-3-prop-2-enylsulfanylpropanoic acid
SMIL N[C@@H](CSCC=C)C(=O)O
Tekniske data
Kemisk navn eller materiale S-Allyl-L-cysteine
InChI formel InChI=1S/C6H11NO2S/c1-2-3-10-4-5(7)6(8)9/h2,5H,1,3-4,7H2,(H,8,9)/t5-/m0/s1
Mængde 10 mg
Formel vægt 161.051