Få mere at vide
L-Cysteinesulfinic Acid, Mikromol™
Artikelnummer. 30050787
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050787 25 mg
30051862 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050787 Leverandør Mikromol Leverandørnr. MM0236.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
L-Cysteinesulfinic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Cysteinesulfinic Acid
CAS 1115-65-7
Analyt- eller komponentnavne L-Cysteinesulfinic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C3 H7 N O4 S
Certificeringer/compliance ISO 17025
Synonym L-Cysteinesulfinic Acid
InChI formel InChI=1S/C3H7NO4S/c4-2(3(5)6)1-9(7)8/h2H,1,4H2,(H,5,6)(H,7,8)/t2-/m0/s1
SMIL N[C@@H](CS(=O)O)C(=O)O
IUPAC navn (2R)-2-amino-3-sulfinopropanoic acid
Molekylvægt (g/mol) 153.16
Mængde 25 mg
Formel vægt 153.0096 g/mol