Få mere at vide
L-Cysteine Hydrochloride Monohydrate, MP Biomedicals
Artikelnummer. 11493400 Shop alle MP Biomedicals produkter
Skift visning
Mængde:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
11493400 500 g
11473400 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11493400 Leverandør MP Biomedicals Leverandørnr. 02101446.5
  • L-Cysteine hydrochloride monohydrate has been used in a protocol for the separation of dorsal root ganglion neurons (DRGs)
  • It has also been used in a study to examine its effect as an inhibitor in preventing enzymatic browning in apples
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
Molekylær formel C3H10ClNO3S
Molekylvægt (g/mol) 175.63
MDL nummer MFCD00065606
InChI nøgle QIJRTFXNRTXDIP-JIZZDEOASA-N
Tekniske data
Kemisk navn eller materiale L-CYSTEINE HYDROCHLORIDE MONOHYDRATE
CAS 1880366
Smeltepunkt 175°C (literature)
Lineær formel C3H7NO2S·HCl· H2O
Mængde 500 g
Synonym 2-propoxy-benzaldehyde, 2-propyloxybenzaldehyde, 2-n-propoxybenzaldehyde, propoxy benzaldehyde, acmc-20anok, pubchem3160, benzaldehyde,-2-propoxy, benzaldehyde,2-propoxy, aronis23744, 2,6-dichloro-4-nitropyridine 3-oxide
Opløselighedsinformation Freely soluble in water (50mg/mL).
SMIL [H+].O.[Cl-].N[C@@H](CS)C(O)=O
IUPAC navn hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride
PubChem CID 249807
Formel vægt 175.64
Fysisk form Krystaller