Få mere at vide
L-Cysteine-D-penicillamine Disulfide, TRC
Artikelnummer. 30361752
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361752 250 mg
30363123 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30361752 Leverandør Toronto Research Chemicals Leverandørnr. TRCC996800250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18840-45-4
Molekylær formel C8H16N2O4S2
Molekylvægt (g/mol) 268.35
Synonym 3-[[(2R)-2-Amino-2-carboxyethyl]dithio]-D-valine, 3,3-Dimethyl-3,3'-dithiodialanine, (R)-3-[(2-Amino-2-carboxyethyl)dithio]-D-valine, 3-[(L-2-Amino-2-carboxyethyl)dithio]-D-valine, Penicillamine Cysteine Disulfide
IUPAC navn (2S)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]-3-methylbutanoic acid
SMIL CC(C)([C@H](C(=O)O)N)SSC[C@@H](C(=O)O)N
Tekniske data
Kemisk navn eller materiale L-Cysteine-D-penicillamine Disulfide
InChI formel InChI=1S/C8H16N2O4S2/c1-8(2,5(10)7(13)14)16-15-3-4(9)6(11)12/h4-5H,3,9-10H2,1-2H3,(H,11,12)(H,13,14)/t4-,5-/m0/s1
Mængde 250 mg
Formel vægt 268.0551