Få mere at vide
L-Cysteine-15N,d3, TRC
Artikelnummer. 30359030
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30359030 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359030 Leverandør Toronto Research Chemicals Leverandørnr. TRCC99500225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Cysteine-15N,d3
CAS 1795787-05-1
Molekylær formel C3 D3 H4 15N O2 S
Mængde 25 mg
Synonym (R)-2-Amino-3-mercaptopropanoic Acid-15N,d3, (R)-Cysteine-15N,d3, 2-Amino-3-mercaptopropionic Acid-15N,d3, Cystein-15N,d3, Cysteine-15N,d3, E 920-15N,d3, Half-cystine-15N,d3, L-(+)-Cysteine-15N,d3, 3-Mercapto-L-alanine-15N,d3, L-Cys-15N,d3, NSC 8746-15N,d3, Thioserine-15N,d3, β-Mercaptoalanine-15N,d3
InChI formel InChI=1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1/i1D2,2D,4+1
SMIL [2H]C([2H])(S)[C@]([2H])([15NH2])C(=O)O
IUPAC navn (2R)-2-(^{15}N)azanyl-2,3,3-trideuterio-3-sulfanyl-propanoic acid
Molekylvægt (g/mol) 125.17
Formel vægt 125.0356