Få mere at vide
L-Cysteine-13C3, TRC
Artikelnummer. 30132924
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30132924 1 mg -
30136095 5 mg -
30012918 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132924 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9950041MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202114-66-7
Molekylær formel 13C H7 N O2 S
Molekylvægt (g/mol) 124.14
Synonym L-Cysteine-1,2,3-13C3
IUPAC navn (2R)-2-amino-3-sulfanyl-(1,2,3-^{13}C_{3})propanoic acid
SMIL N[13C@@H]([13CH2]S)[13C](=O)O
Tekniske data
Kemisk navn eller materiale L-Cysteine-13C3
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1/i1+1,2+1,3+1
Formel vægt 124.0298
Procent renhed >95%