missing translation for 'onlineSavingsMsg'
Få mere at vide

L-Cystathionine, MedChemExpress

Artikelnummer. 16412678
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16412678 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16412678 Leverandør Medchem Express Leverandørnr. HYW009749

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress L-Cystathionine is a nonprotein thioether and is a key amino acid associated with the metabolic state of sulfur-containing amino acids. L-Cystathionine protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs). L-Cystathionine plays an important role in cardiovascular protection.

L-Cystathionine is a nonprotein thioether and is a key amino acid associated with the metabolic state of sulfur-containing amino acids. L-Cystathionine protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs). L-Cystathionine plays an important role in cardiovascular protection.

Tekniske data

Kemisk navn eller materiale L-Cystathionine
CAS 56-88-2
Farve White
Renhedsgrad noter Research
Molekylær formel C7H14N2O4S
Mængde 5 mg
Opløselighedsinformation 1M HCl : 50 mg/mL (224.96 mM; ultrasonic and adjust pH to 1 with HCl) ∣H2O : 25 mg/mL (112.48 mM; ultrasonic and warming and adjust pH to 11 with NaOH and heat to 80°C) ∣DMSO : 1 mg/mL (4.50 mM; Need ultrasonic)
SMIL N[C@@H](CCSC[C@H](N)C(O)=O)C(O)=O
Molekylvægt (g/mol) 222.26
Formel vægt 222.26
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Metabolism-protein/nucleotide metabolism
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.