Få mere at vide
L-Carnitine-d9, TRC
Artikelnummer. 30481600
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30481600 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481600 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1841115MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Carnitine trimethyl-d9
CAS 126827-79-0
Molekylær formel C7 D9 H7 N O3
Mængde 5 mg
Synonym 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-tri(methyl-d3)-, inner salt, (2R)- (9CI), 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-tri(methyl-d3)-, inner salt, (R)- (ZCI), (2R)-3-Carboxy-2-hydroxy-N,N,N-tri(methyl-d3)-1-propanaminium inner salt, L-Carnitine-d9, L-Carnitine-D9 (N,N,N-trimethyl-D9)
InChI formel InChI=1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/p+1/t6-/m1/s1/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])[N+](C[C@H](O)CC(=O)O)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn [(2R)-3-carboxy-2-hydroxy-propyl]-tris(trideuteriomethyl)ammonium
Molekylvægt (g/mol) 171.26
Formel vægt 171.169