Få mere at vide
L-Carnitine-d3 Chloride, TRC
Artikelnummer. 30364919
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30364919 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30364919 Leverandør Toronto Research Chemicals Leverandørnr. TRCC184113100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Carnitine-d3 Chloride
CAS 350818-62-1
Renhedsgrad noter HPLC
Molekylær formel C7 D3 H13 N O3 . Cl
Mængde 100 mg
Synonym 1-Propanaminium, 3-carboxy-2-hydroxy-N,N-dimethyl-N-(methyl-d3)-, chloride, (2R)- (9CI), (2R)-3-Carboxy-2-hydroxy-N,N-dimethyl-N-(methyl-d3)-1-propanaminium chloride, (R)-(3-Carboxy-2-hydroxypropyl)dimethyl(methyl-d3)ammonium chloride, (R)-Carnitine-d3 hydrochloride, Levocarnitine-d3 chloride, l-Carnitine-d3 chloride, l-Carnitine-d hydrochloride, Levocarnitine-D3 (N-methyl-D3) chloride
InChI formel InChI=1S/C7H15NO3.ClH/c1-8(2,3)5-6(9)4-7(10)11;/h6,9H,4-5H2,1-3H3;1H/t6-;/m1./s1/i1D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@H](O)CC(=O)O
IUPAC navn [(2R)-3-carboxy-2-hydroxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 200.678
Formel vægt 200.101
Procent renhed >95