Få mere at vide
L-BMAA hydrochloride, R&D Systems™
Beskrivelse
Neurotoxic glutamate agonist originally isolated from Cycas circinalis. Implicated in the pathogenesis of amyotrophic lateral sclerosis-Parkinsonism-dementia complex of Guam (Guam ALS-PD).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-BMAA hydrochloride |
| CAS | 16012-55-8 |
| Molekylær formel | C4H11ClN2O2 |
| Synonym | l-bmaa hydrochloride, s-2-amino-3-methylamino propanoic acid hydrochloride, beta-n-methylamino-l-alanine hydrochloride, s +-2-amino-3-methylamino propionic acid hydrochloride, s-2-amino-3-methylamino-propionic acid hydrochloride, l-2-amino-3 methylamino propanoicacidhydrochloride, h-l-dap me-oh hcl, s +-2-amino-3-methylamino propionicacidhydrochloride, bestipharma 572-892, beta-methylamino-l-alanine hcl |
| InChI nøgle | VDXYGASOGLSIDM-DFWYDOINSA-N |
| SMIL | CNCC(C(=O)O)N.Cl |
| IUPAC navn | (2S)-2-amino-3-(methylamino)propanoic acid;hydrochloride |
| Molekylvægt (g/mol) | 154.594 |
| Mængde | 10 mg |
| PubChem CID | 51358333 |
| Vis mere |