missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Aspartic Acid Sodium Salt Monohydrate, ≥98% (dry basis), Thermo Scientific™
Click to view available options
Quantity:
1 kg
10 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Linear Formula | NH2CH(COOH)CH2COONa·H2O |
Formula Weight | 173.1 |
Packaging | Plastic bottle |
Chemical Name or Material | L to Aspartic Acid Sodium Salt Monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'