missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%

Brand:  Alfa Aesar™ H63257.MD

Additional Details : CAS Number : 2673-19-0 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15488196

  • 211.06 DKK / 250mg


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

2673-19-0
239.696
SFYKWYAIJZEDNG-RGMNGODLSA-N
13970777
CC(C)(C)OC(=O)CC(C(=O)OC)N.Cl
C9H18ClNO4
MFCD00153427
h-asp otbu-ome.hcl, h-asp otbu-ome hcl, l-aspartic acid 4-tert-butyl-1-methyl ester hydrochloride, h-asp otbu-ome hydrochloride, h-asp obut-ome hcl, s-4-tert-butyl 1-methyl 2-aminosuccinate hydrochloride, 4-tert-butyl 1-methyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-ome, h-asp otbu-omecl, h-asp o-tbu-ome
4-O-tert-butyl 1-O-methyl (2S)-2-aminobutanedioate;hydrochloride

Specifications

L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride
C9H18ClNO4
6005591
SFYKWYAIJZEDNG-RGMNGODLSA-N
4-O-tert-butyl 1-O-methyl (2S)-2-aminobutanedioate;hydrochloride
13970777
Solid
250mg
2673-19-0
MFCD00153427
h-asp otbu-ome.hcl, h-asp otbu-ome hcl, l-aspartic acid 4-tert-butyl-1-methyl ester hydrochloride, h-asp otbu-ome hydrochloride, h-asp obut-ome hcl, s-4-tert-butyl 1-methyl 2-aminosuccinate hydrochloride, 4-tert-butyl 1-methyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-ome, h-asp otbu-omecl, h-asp o-tbu-ome
CC(C)(C)OC(=O)CC(C(=O)OC)N.Cl
239.696
239.7
98%
Videos
SDS Documents