Få mere at vide
L-Aspartic Acid-15N, TRC
Artikelnummer. 30135865
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135865 50 mg
30134958 100 mg
30006761 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135865 Leverandør Toronto Research Chemicals Leverandørnr. TRCA79002250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3715-16-0
Molekylær formel C4 H7 15N O4
Molekylvægt (g/mol) 134.096
Synonym (S)-2-Aminobutanedioic Acid-15N, (S)-Aminobutanedioic Acid-15N, L-(+)-Aspartic Acid-15N, L-Aminosuccinic Acid-15N, L-Asparagic Acid-15N, L-Asparaginic Acid-15N, NSC 3973-15N, NSC 79553-15N
IUPAC navn (2S)-2-(^{15}N)azanylbutanedioic acid
SMIL [15NH2][C@@H](CC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Aspartic Acid-15N
Mængde 50 mg
InChI formel InChI=1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1/i5+1
Formel vægt 134.0345