Få mere at vide
L-Asparagine-13C4, 15N2 Hydrate, TRC
Artikelnummer. 30135619
Skift visning
Mængde:
0.25 mg
0.5 mg
2.5 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135619 0.25 mg -
30134285 0.5 mg 0.5mg
30007516 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135619 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7900380.25MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202406-87-9
Molekylær formel 13C4H815N2O3 H2O
Molekylvægt (g/mol) 138.081802
Synonym L-Asparagine-13C4-15N2, monohydrate (9CI), L-Asparagine-1,2,3,4-13C4-N,N2-15N2 hydrate (1:1), (2S)-2,4-Di(amino-15N)-4-oxobutanoic-1,2,3,4-13C4 acid hydrate (1:1), (S)-2,4-Diamino-4-oxobutanoic acid-13C4,15N2 monohydrate, L-Asparagine-13C4,15N2 monohydrate
SMIL O=[13C](O)[13C@@H]([15NH2])[13CH2][13C]([15NH2])=O.O
Tekniske data
Kemisk navn eller materiale L-Asparagine-13C4, 15N2 Hydrate
Renhedsgrad noter HPLC
Mængde 0.25 mg
Procent renhed >95%