missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Ascorbic acid, Electrophoresis Grade, 99+%, Thermo Scientific Chemicals

Catalog Number p-7057473
Click to view available options
Quantity:
50 g
100 g
This item is not returnable. View return policy
50-81-7
C6H8O6
176.12
MFCD00064328
CIWBSHSKHKDKBQ-DOMZIZNONA-N
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
54670067
CHEBI:29073
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
This item is not returnable. View return policy
50-81-7
C6H8O6
176.12
MFCD00064328
CIWBSHSKHKDKBQ-DOMZIZNONA-N
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
54670067
CHEBI:29073
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 190°C to 193°C (decomposition)
Color White
Density 1.65 g/mL
Quantity 50 g
Beilstein 84272
Merck Index 14,830
Solubility Information Soluble in water at 0;5 M
Formula Weight 176.12
Percent Purity ≥99%
Grade Electrophoresis
Physical Form Powder
Chemical Name or Material L-(+)-Ascorbic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'