Få mere at vide
L-Arginine Ethyl Ester Dihydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Arginine Ethyl Ester Dihydrochloride |
| CAS | 36589-29-4 |
| Molekylær formel | C8 H18 N4 O2 . 2 Cl H |
| Mængde | 10 g |
| Synonym | L-Arginine, ethyl ester, dihydrochloride (9CI), Arginine ethyl ester dihydrochloride, L-Arginine, ethyl ester, hydrochloride (1:2), L-Arginine Ethyl Ester Dihydrochloride |
| InChI formel | InChI=1S/C8H18N4O2.2ClH/c1-2-14-7(13)6(9)4-3-5-12-8(10)11;;/h6H,2-5,9H2,1H3,(H4,10,11,12);2*1H/t6-;;/m0../s1 |
| SMIL | Cl.Cl.CCOC(=O)[C@@H](N)CCCNC(=N)N |
| IUPAC navn | ethyl (2S)-2-amino-5-carbamimidamidopentanoate;dihydrochloride |
| Molekylvægt (g/mol) | 275.18 |
| Formel vægt | 274.0963 |