Få mere at vide
L-Anserine-d4 (N-b-alanyl-d4), TRC
Artikelnummer. 30334257
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30334257 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334257 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6782542.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Anserine-d4 (N-beta-alanyl-d4)
CAS 1201658-81-2
Molekylær formel C10 D4 H12 N4 O3
Mængde 2.5 mg
Synonym L-Anserine-d4 (N-β-alanyl-d4), L-Histidine, β-alanyl-2,2,3,3-d4-3-methyl- (ACI), β-Alanyl-2,2,3,3-d4-3-methyl-L-histidine (ACI)
InChI formel InChI=1S/C10H16N4O3/c1-14-6-12-5-7(14)4-8(10(16)17)13-9(15)2-3-11/h5-6,8H,2-4,11H2,1H3,(H,13,15)(H,16,17)/t8-/m0/s1/i2D2,3D2
SMIL [2H]C([2H])(N)C([2H])([2H])C(=O)N[C@@H](Cc1cncn1C)C(=O)O
IUPAC navn (2S)-2-[(3-amino-2,2,3,3-tetradeuteriopropanoyl)amino]-3-(3-methylimidazol-4-yl)propanoic acid
Molekylvægt (g/mol) 244.29
Formel vægt 244.1473