Få mere at vide
L-(+)-Ampicillin, TRC
Artikelnummer. 30136952
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136952 50 mg
30135741 10 mg
30007287 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136952 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63433550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19379-33-0
Molekylær formel C16 H19 N3 O4 S
Molekylvægt (g/mol) 349.4
Synonym (2S,5R,6R)-6-[[(2S)-2-Amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, Ampicillin Sodium Imp. B (EP), L-Ampicillin
IUPAC navn (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMIL CC1(C)S[C@@H]2[C@H](NC(=O)[C@@H](N)c3ccccc3)C(=O)N2[C@H]1C(=O)O
Tekniske data
Kemisk navn eller materiale (2S,5R,6R)-6-[((2S)-2-Amino-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (L-Ampicillin)
Mængde 50 mg
InChI formel InChI=1S/C16H19N3O4S/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23)/t9-,10+,11-,14+/m0/s1
Formel vægt 349.1096