missing translation for 'onlineSavingsMsg'
Få mere at vide

L-Alpha-Methyl DOPA Methyl Ester Hydrochloride, TRC

Artikelnummer. 30476125
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476125 1 g
30473392 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30476125 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3038101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115054-62-1
Molekylær formel C11 H15 N O4 . Cl H
Molekylvægt (g/mol) 261.7
Synonym L-Tyrosine, 3-hydroxy-α-methyl-, methyl ester, hydrochloride (1:1), L-Tyrosine, 3-hydroxy-α-methyl-, methyl ester, hydrochloride (9CI), L-α-Methyldopa methyl ester hydrochloride, Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride, Methyldopa Methyl Ester Hydrochloride
IUPAC navn methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate;hydrochloride
SMIL Cl.COC(=O)[C@@](C)(N)Cc1ccc(O)c(O)c1

Specifications

Kemisk navn eller materiale Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride
Mængde 1 g
InChI formel InChI=1S/C11H15NO4.ClH/c1-11(12,10(15)16-2)6-7-3-4-8(13)9(14)5-7;/h3-5,13-14H,6,12H2,1-2H3;1H/t11-;/m0./s1
Formel vægt 261.0768
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.